Aluminium oxide activated neutral Brockmann Grade II

Supplier: Thermo Scientific Chemicals

A17141-0E A17141-36
AAAA17141-0EEA 210 USD
AAAA17141-0E AAAA17141-36
Aluminium oxide activated neutral Brockmann Grade II
Aluminium oxide activated neutral

CAS No.: 1344-28-1 Molecular Formula: Al2O3 Formula Weight: 101.96 Flash Point: None Odor: Odorless MDL No.: MFCD00003424

Formula: Al₂O₃
Density: 3.97
Storage Temperature: Ambient
MDL Number: MFCD00003424
CAS Number: 1344-28-1
EINECS: 215-691-6
Merck Index: 14,356

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR