N,N'-Diisopropylethylenediamine 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
N,N’-Diisopropylethylenediamine
A17114-22
A17114-14
AAAA17114-22EA
690
USD
AAAA17114-22
AAA17114-14
N,N'-Diisopropylethylenediamine 97%
N,N'-Diisopropylethylenediamine
Purity: 97%
CAS No.: 4013-94-9
Molecular Formula: C8H20N2
Formula Weight: 144.26
Flash Point: 51°(123°F)
Refractive Index: 1.4289
Boiling Point: 169-171°
Beilstein Reference: 1735571
UN No.: 2733
MDL No.: MFCD00008864
Formula:
C₈H₂₀N₂ MW: 144.26 g/mol Boiling Pt: 169…171 °C Density: 0.798 Flash Pt: 51 °C(123 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008864 CAS Number: 4013-94-9 EINECS: 223-666-6 UN: 2734 ADR: "8",II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...