2-(1-Naphthyl)-5-phenyloxazole ≥99% laser grade, for scintillation

Supplier: Thermo Scientific Chemicals

Synonyms: ANPO

A16679-03 A16679-06
AAA16679-03EA 76.6 USD
AAA16679-03 AAA16679-06
2-(1-Naphthyl)-5-phenyloxazole ≥99% laser grade, for scintillation
2-(1-Naphthyl)-5-phenyloxazole

Purity: 99+% CAS No.: 846-63-9 Molecular Formula: C19H13NO Formula Weight: 271.32 Melting Point: 104-105° Beilstein Reference: 210666 MDL No.: MFCD00019714

Formula: C₁₉H₁₃NO
MW: 271.32 g/mol
Melting Pt: 104…105 °C
Storage Temperature: Ambient
MDL Number: MFCD00019714
CAS Number: 846-63-9
EINECS: 212-689-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR