2',7'-Dichlorofluorescein

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2',7'-Dichloro-3',6'-dihydroxy-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one

A16212-06 A16212-14
AAA16212-06EA 93.43 USD
AAA16212-06 AAA16212-14
2',7'-Dichlorofluorescein
2',7'-Dichlorofluorescein

As a fluorescent indicator


CAS No.: 76-54-0 Molecular Formula: C20H10Cl2O5 Formula Weight: 401.20 Melting Point: 300° dec. MDL No.: MFCD00005047

Formula: C₂₀H₁₀Cl₂O₅
MW: 401.20 g/mol
Melting Pt: >300 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00005047
CAS Number: 76-54-0
EINECS: 200-968-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR