Heparin sodium salt (from porcine intestinal mucosa), IU>=100/mg

Supplier: Thermo Scientific Chemicals

Synonyms: Reviparin sodium

A16198-03 A16198-06 A16198-MD
AAA16198-03EA 243.68 USD
AAA16198-03 AAA16198-06 AAAA16198-MD
Heparin sodium salt (from porcine intestinal mucosa), IU>=100/mg
Heparin sodium salt

Blood anti-coagulant


Applications: Blood anti-coagulant


Applications: Blood anti-coagulant

Storage Temperature: Ambient
MDL Number: MFCD00081689
CAS Number: 9041-08-1
Merck Index: 14,4653

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR