meso-Erythritol 99%
Supplier: Thermo Scientific Chemicals
A15813-22
A15813-14
AAA15813-22EA
140
USD
AAA15813-22
AAAA15813-14
meso-Erythritol 99%
meso-Erythritol
Purity: 99%
CAS No.: 149-32-6
Molecular Formula: C4H10O4
Formula Weight: 122.12
Melting Point: 119-124°
Boiling Point: 329-331°
Beilstein Reference: 1719753
MDL No.: MFCD00004710
Formula:
C₄H₁₀O₄ Boiling Pt: 329…331 °C Melting Pt: 119…124 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004710 CAS Number: 149-32-6 EINECS: 205-737-3 Merck Index: 14,3675 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...