Allantoin 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
1-(2,5-Dioxoimidazolidin-4-yl)urea
A15571-0B
A15571-0I
A15571-30
AAA15571-0BEA
156
USD
AAA15571-0B
AAA15571-0I
AAA15571-30
Allantoin 98%
Allantoin
Purity: 98%
CAS No.: 97-59-6
Molecular Formula: C4H6N4O3
Formula Weight: 158.12
Melting Point: ca 230° dec.
Beilstein Reference: 102364
Merck Reference: 14,258
MDL No.: MFCD00005260
Formula:
C₄H₆N₄O₃ MW: 158.12 Melting Pt: ca 230 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00005260 CAS Number: 97-59-6 EINECS: 202-592-8 Merck Index: 14,258 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...