3,4-Diaminoanisole 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Methoxy-o-phenylenediamine
A15557-03
A15557-06
A15557-14
AAA15557-03EA
49.4
USD
AAA15557-03
AAA15557-06
AAA15557-14
3,4-Diaminoanisole 98%
3,4-Diaminoanisole
Purity: 98%
CAS No.: 102-51-2
Molecular Formula: C7H10N2O
Formula Weight: 138.17
Storage Temperature: 2°C to 8°C
Melting Point: 46-50°
UN No.: 2811
MDL No.: MFCD00047837
Formula:
C₇H₁₀N₂O MW: 138.17 g/mol Melting Pt: 46…50 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00047837 CAS Number: 102-51-2 UN: 2811 ADR: "6.1",III |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...