Metaldehyde 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A15292-18
A15292-30
A15292-0B
AAA15292-18EA
29.9
USD
AAA15292-18
AAA15292-30
AAA15292-0B
Metaldehyde 98%
Metaldehyde
Purity: 98%
CAS No.: 108-62-3
Molecular Formula: C8H16O4
Formula Weight: 176.22
Flash Point: 55°(131°F)
Melting Point: 192-196° subl.
UN No.: 1332
MDL No.: MFCD00071549
Formula:
C₈H₁₆O₄ Melting Pt: 192…196 °C (subl.) Flash Pt: 55 °C(131 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00071549 CAS Number: 108-62-3 EINECS: 203-600-2 UN: 1332 ADR: "4.1",III Merck Index: 14,5923 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...