2,3-Diphenylpyrazine 98%

Supplier: Thermo Scientific Chemicals

A14700-06 A14700-03
AAA14700-06EA 129 USD
AAA14700-06 AAA14700-03
2,3-Diphenylpyrazine 98%
2,3-Diphenylpyrazine

Purity: 98% CAS No.: 1588-89-2 Molecular Formula: C16H12N2 Formula Weight: 232.29 Odor: Odorless Melting Point: 119-122° MDL No.: MFCD00053000

Formula: C₁₆H₁₂N₂
MW: 232.29 g/mol
Melting Pt: 119…122 °C
Storage Temperature: Ambient
MDL Number: MFCD00053000
CAS Number: 1588-89-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR