Geranyl bromide 96%

Supplier: Thermo Scientific Chemicals
Danger

A14093-06 A14093-22 A14093-14
AAA14093-06EA 59.3 USD
AAA14093-06 AAA14093-22 AAA14093-14
Geranyl bromide 96%
Geranyl bromide

Purity: 96% CAS No.: 6138-90-5 Molecular Formula: C10H17Br Formula Weight: 217.16 Storage Temperature: 2°C to 8°C Flash Point: 95°(203°F) Refractive Index: 1.5040 Boiling Point: 99-101°/10mm Beilstein Reference: 1422412 MDL No.: MFCD00000243

Formula: C₁₀H₁₇Br
MW: 217.16 g/mol
Boiling Pt: 99…101 °C/10 mm
Density: 1.094
Flash Pt: 95 °C(203 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00000243
CAS Number: 6138-90-5
EINECS: 228-123-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR