2-Vinylpyrazine 98% stabilized
Supplier: Thermo Scientific Chemicals
|
Warning
|
Stabilization:
Stabilised with hydroquinone (HQ) ∼0.1%
A14007-14
A14007-03
A14007-06
AAAA14007-14EA
835
USD
AAAA14007-14
AAAA14007-03
AAAA14007-06
2-Vinylpyrazine 98% stabilized
2-Vinylpyrazine
Purity: 98%
CAS No.: 4177-16-6
Molecular Formula: C6H6N2
Formula Weight: 106.13
Storage Temperature: -30°C to -10°C
Flash Point: 60°(140°F)
Refractive Index: 1.5560
Boiling Point: 163-164°
Beilstein Reference: 105566
UN No.: 1993
MDL No.: MFCD00051591
Formula:
C₆H₆N₂ Boiling Pt: 163…164 °C Density: 1.041 Flash Pt: 60 °C(140 °F) Storage Temperature: Freezer |
MDL Number:
MFCD00051591 CAS Number: 4177-16-6 EINECS: 224-045-2 UN: 1993 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...