EEDQ (ethyl-N-(2-ethoxy-1,2-dihydroquinoline)carboxylate) 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Ethoxy-1-ethoxycarbonyl-1,2-dihydroquinoline, N-Ethoxycarbonyl-2-ethoxy-1,2-dihydroquinoline, Ethyl 2-ethoxyquinoline-1(2H)-carboxylate

A13724-18 A13724-09 A13724-30
AAA13724-18EA 96.9 USD
AAA13724-18 AAA13724-09 AAA13724-30
EEDQ (ethyl-N-(2-ethoxy-1,2-dihydroquinoline)carboxylate) 99%
EEDQ (ethyl-N-(2-ethoxy-1,2-dihydroquinoline)carboxylate)

Fieser: 2,191 3,137 4,223 21,185


Purity: 99% CAS No.: 16357-59-8 Molecular Formula: C14H17NO3 Formula Weight: 247.29 Storage Temperature: 2°C to 8°C Melting Point: 63-66° Boiling Point: 125-128°/0.1mm Beilstein Reference: 533048 Merck Reference: 14,3518 MDL No.: MFCD00006703

Formula: C₁₄H₁₇NO₃
MW: 247.29 g/mol
Boiling Pt: 125…128 °C/0.1 mm
Melting Pt: 63…66 °C
Storage Temperature: Ambient
MDL Number: MFCD00006703
CAS Number: 16357-59-8
EINECS: 240-418-2
Merck Index: 14,3518

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR