Diethyl-bis(hydroxymethyl)malonate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Diethyl 2,2-bis(hydroxymethyl)malonate

A13677-14 A13677-22
AAA13677-14EA 78.5 USD
AAA13677-14 AAA13677-36 AAA13677-22
Diethyl-bis(hydroxymethyl)malonate 97%
Diethyl-bis(hydroxymethyl)malonate

Purity: 97% CAS No.: 20605-01-0 Molecular Formula: C9H16O6 Formula Weight: 220.22 Flash Point: 110°(230°F) Melting Point: 47-52° Beilstein Reference: 1788515 MDL No.: MFCD00009130

Formula: C₉H₁₆O₆
MW: 220.221 g/mol
Melting Pt: 47…52 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009130
CAS Number: 20605-01-0
EINECS: 243-916-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR