2-Phenylpropionitrile 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: alpha-Methylphenylacetonitrile

A13636-14 A13636-09
AAAA13636-14EA 108 USD
AAAA13636-14 AAA13636-09 AAA13636-18
2-Phenylpropionitrile 96%
2-Phenylpropionitrile

Purity: 97% CAS No.: 1823-91-2 Molecular Formula: C9H9N Formula Weight: 131.18 Flash Point: 93°(199°F) Refractive Index: 1.5106 Boiling Point: 230-232° Beilstein Reference: 774576 UN No.: 3276 MDL No.: MFCD00001871

Formula: C₉H₉N
MW: 131.18 g/mol
Boiling Pt: 230…232 °C
Density: 0.985
Flash Pt: 93 °C (199 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001871
CAS Number: 1823-91-2
EINECS: 217-354-9
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR