(+)-Rutin trihydrate 95%
Supplier: Thermo Scientific Chemicals
A13570-22
A13570-14
A13570-36
AAA13570-22EA
93.7
USD
AAA13570-22
AAA13570-14
AAA13570-36
(+)-Rutin trihydrate 95%
(+)-Rutin trihydrate
Purity: 97%
CAS No.: 250249-75-3
Molecular Formula: C27H30O16.3H2O
Formula Weight: 664.58 (610.52anhy)
Melting Point: 184° dec.
Beilstein Reference: 75455
Optical Rotation: +9° (c=0.5 in ethanol)
MDL No.: MFCD00149490
Formula:
C₂₇H₃₀O₁₆·3H₂O MW: 664.58 g/mol (610.52 anhy) Melting Pt: 184 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00149490 CAS Number: 250249-75-3 EINECS: 205-814-1 Merck Index: 14,3802 |
Specification Test Results
Optical rotation | +9° (c=0.5 in ethanol) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...