3-Pyridinesulfonic acid 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Pyridine-3-sulphonic acid

A13101-14 A13101-36 A13101-22
AAA13101-14EA 66.7 USD
AAA13101-14 AAA13101-36 AAA13101-22
3-Pyridinesulfonic acid 98+%
3-Pyridinesulphonic acid

Purity: 98+% CAS No.: 636-73-7 Molecular Formula: C5H5NO3S Formula Weight: 159.16 Melting Point: ca 350° dec. Beilstein Reference: 120822 UN No.: 2585 MDL No.: MFCD00006381

Formula: C₅H₅NO₃S
Melting Pt: ca 350 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00006381
CAS Number: 636-73-7
EINECS: 211-265-9
UN: 2585
ADR: "8",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR