3-Aminopropionitrile 98% stabilized

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: β-Aminopropionitrile, 3-Aminopropanenitrile

Stabilization: Stabilised with potassium carbonate

A13043-03 A13043-06 A13043-14
AAA13043-03EA 44.4 USD
AAA13043-03 AAA13043-06 AAA13043-14
3-Aminopropionitrile 98% stabilized
3-Aminopropionitrile

Purity: 98% CAS No.: 151-18-8 Molecular Formula: C3H6N2 Formula Weight: 70.10 Storage Temperature: 2°C to 8°C Refractive Index: 1.4375 Odor: Amine-like Boiling Point: 79-81°/16mm Beilstein Reference: 1698848 Merck Reference: 14,469 UN No.: 2735 MDL No.: MFCD00014820

Formula: C₃H₆N₂
MW: 70.10 g/mol
Boiling Pt: 79…81 °C/16 mm
Density: 0.952
Storage Temperature: Refrigerator
MDL Number: MFCD00014820
CAS Number: 151-18-8
EINECS: 205-786-0
UN: 2922
ADR: "8",III
Merck Index: 14,469

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR