Ethyl cyano(hydroxyimino)acetate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ethyl 2-cyano-2-(hydroxyimino)acetate, Ethyl Cyanoglyoxalate Oxime, Cyano(hydroxyimino)acetic Acid Ethyl Ester, Oxyma

A12691-06 A12691-22 A12691-14
AAA12691-06EA 45.1 USD
AAA12691-06 AAA12691-22 AAA12691-14
Ethyl cyano(hydroxyimino)acetate 97%
Ethyl cyano(hydroxyimino)acetate

This product is a non-explosive alternative to HOBt or HOAt, and allows high peptide coupling rates at low racemization when applied in combination with carbodiimides.


Purity: 97% CAS No.: 3849-21-6 Molecular Formula: C5H6N2O3 Formula Weight: 142.11 Melting Point: 127-132° MDL No.: MFCD00000625

Formula: C₅H₆N₂O₃
Melting Pt: 127…132 °C
Storage Temperature: Ambient
MDL Number: MFCD00002112
CAS Number: 3849-21-6
EINECS: 223-351-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR