Ethyl hydrogen malonate 96%

Supplier: Thermo Scientific Chemicals
Warning

A12627-09 A12627-18 A12627-30
AAAA12627-09EA 68.4 USD
AAAA12627-09 AAAA12627-18 AAAA12627-30
Ethyl hydrogen malonate 96%
Ethyl hydrogen malonate

Fieser: 6,255 10,182 15,156


Purity: 96% CAS No.: 1071-46-1 Molecular Formula: C5H8O4 Formula Weight: 132.12 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Physical Form: Liquid Density: 1.119 Refractive Index: 1.4295 Appearance: Colorless to yellow Boiling Point: 106°/3mm Beilstein Reference: 1758845 MDL No.: MFCD00020490

Formula: C₅H₈O₄
Boiling Pt: 106 °C (3 mmHg)
Density: 1.119
Flash Pt: >110 °C (230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00020490
CAS Number: 1071-46-1
EINECS: 213-992-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR