4,6-Dimethyl-2-oxo-1,2-dihydropyridine-3-carbonitrile 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12466-14
A12466-22
AAAA12466-06EA
36.73
USD
AAAA12466-06
AAAA12466-14
AAAA12466-22
4,6-Dimethyl-2-oxo-1,2-dihydropyridine-3-carbonitrile 98%
4,6-Dimethyl-2-oxo-1,2-dihydropyridine-3-carbonitrile
Purity: 98%
CAS No.: 769-28-8
Molecular Formula: C8H8N2O
Formula Weight: 148.17
Melting Point: ca 290° dec.
Beilstein Reference: 130992
MDL No.: MFCD00006269
Formula:
C₈H₈N₂O Melting Pt: ∼ 290 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00006269 CAS Number: 769-28-8 EINECS: 212-207-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...