Trimethyl-4-phosphonocrotonate mixture E/Z ≥90%

Supplier: Thermo Scientific Chemicals
Warning

A12399-18 A12399-09
AAA12399-30EA 1521.66 USD
AAA12399-30 AAA12399-18 AAAA12399-09
Trimethyl-4-phosphonocrotonate mixture E/Z ≥90%
Trimethyl-4-phosphonocrotonate

Purity: 90+% CAS No.: 86120-40-3 Molecular Formula: C7H13O5P Formula Weight: 208.15 Boiling Point: 123-124°/1mm Beilstein Reference: 4987133 MDL No.: MFCD00051564

Formula: C₇H₁₃O₅P
MW: 208.15 g/mol
Boiling Pt: 123…124 °C/1 mm
Storage Temperature: Ambient
MDL Number: MFCD00051564
CAS Number: 86120-40-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR