Phosphoenolpyruvic acid monocyclohexylammonium salt 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Phosphoenolpyruvic acid cyclohexylammonium salt, PEP·CHX, Phospho(enol)pyruvic acid cyclohexylammonium salt, 2-(Phosphonooxy)-2-propenoic acid monocyclohexylammonium salt

A12385-14 A12385-03 A12385-06
AAA12385-14EA 1207 USD
AAA12385-14 AAA12385-03 AAAA12385-06
Phosphoenolpyruvic acid monocyclohexylammonium salt 98%
Phosphoenolpyruvic acid monocyclohexylammonium salt

Purity: 98% CAS No.: 10526-80-4 Molecular Formula: C9H18NO6P Formula Weight: 267.22 Storage Temperature: -30°C to -10°C Melting Point: ca 140° dec. MDL No.: MFCD00036375

Formula: C₃H₅O₆P
MW: 267.22 g/mol
Melting Pt: ∼ 140 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00036375
CAS Number: 10526-80-4
EINECS: 234-084-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR