3'-Acetamidoacetophenone 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Acetamidoacetophenone

A12188-22 A12188-14
AAA12188-22EA 200 USD
AAA12188-22 AAA12188-14
3'-Acetamidoacetophenone 98%
3'-Acetamidoacetophenone

Purity: 98% CAS No.: 7463-31-2 Molecular Formula: C10H11NO2 Formula Weight: 177.20 Melting Point: 125-129° Beilstein Reference: 2363443

Formula: C₁₀H₁₁NO₂
Melting Pt: 125…129 °C
Storage Temperature: Ambient
MDL Number: MFCD00032278
CAS Number: 7463-31-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR