(Benzoylmethylene)triphenylphosphorane 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Phenyl-2-(triphenylphosphoranylidene)ethanone

A12184-22 A12184-06 A12184-14
AAA12184-22EA 499 USD
AAA12184-22 AAA12184-06 AAA12184-14
(Benzoylmethylene)triphenylphosphorane 98+%
(Benzoylmethylene)triphenylphosphorane

Purity: 98+% CAS No.: 859-65-4 Molecular Formula: C26H21OP Formula Weight: 380.43 Storage Temperature: 2°C to 8°C Melting Point: 183-186° Beilstein Reference: 620285 MDL No.: MFCD00014088

Formula: C₂₆H₂₁OP
MW: 380.43 g/mol
Melting Pt: 183…186 °C
Storage Temperature: Ambient
MDL Number: MFCD00014088
CAS Number: 859-65-4
EINECS: 212-727-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR