3-(4-Hydroxy-3-methoxyphenyl)propionic acid 97%
Supplier: Thermo Scientific Chemicals
|
|
Synonyms:
Dihydroferulic acid, 3-(4-Hydroxy-3-methoxyphenyl)propanoic acid
A12069-06
A12069-03
AAA12069-06EA
308
USD
AAA12069-06
AAA12069-03
3-(4-Hydroxy-3-methoxyphenyl)propionic acid 97%
3-(4-Hydroxy-3-methoxyphenyl)propionic acid
Purity: 97%
CAS No.: 1135-23-5
Molecular Formula: C10H12O4
Formula Weight: 196.20
Melting Point: 87-92°
Beilstein Reference: 2110370
MDL No.: MFCD00016558
Formula:
C₁₀H₁₂O₄ MW: 196.20 g/mol Melting Pt: 87…92 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016558 CAS Number: 1135-23-5 EINECS: 214-489-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...