3,4,5-Tribromo-1H-pyrazole 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3,4,5-Tribromopyrazole
A12031-14
A12031-06
AAA12031-14EA
158
USD
AAA12031-14
AAAA12031-22
AAA12031-06
3,4,5-Tribromo-1H-pyrazole 97%
3,4,5-Tribromo-1H-pyrazole
Purity: 97%
CAS No.: 17635-44-8
Molecular Formula: C3HBr3N2
Formula Weight: 304.78
Odor: Odorless
Melting Point: 184-188°
Beilstein Reference: 113501
MDL No.: MFCD00040248
Formula:
C₃HBr₃N₂ MW: 304.78 g/mol Melting Pt: 184…188 °C Storage Temperature: Ambient |
MDL Number:
MFCD00040248 CAS Number: 17635-44-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...