p-Toluenesulfinic Acid Sodium Salt, (max. 5% H₂O) 97% (dry weight)

Supplier: Thermo Scientific Chemicals

Synonyms: Sodium 4-methylbenzenesulphinate, Sodium-p-toluenesulphinate, 4-Methylbenzenesulphinic acid sodium salt, Sodiumtoluene-4-sulphinate

A11430-36 A11430-22 A11430-14
AAA11430-36EA 132 USD
AAA11430-36 AAA11430-22 AAAA11430-14
p-Toluenesulfinic Acid Sodium Salt, (max. 5% H₂O) 97% (dry weight)
p-Toluenesulfinic Acid Sodium Salt

Fieser: 7,374 8,488 18,324


Purity: 97% CAS No.: 824-79-3 Molecular Formula: C7H7NaO2S Formula Weight: 178.19 Melting Point: 300° Beilstein Reference: 4621454 MDL No.: MFCD00149640

Formula: C₇H₈O₂S
MW: 178.19 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00149640
CAS Number: 824-79-3
EINECS: 212-538-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR