2-(1,3-Benzothiazol-2-yl)acetonitrile 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-(Benzo[d]thiazol-2-yl)acetonitrile

A11343-06 A11343-14
AAA11343-06EA 97.5 USD
AAA11343-06 AAA11343-14
2-(1,3-Benzothiazol-2-yl)acetonitrile 98%
2-(1,3-Benzothiazol-2-yl)acetonitrile

Purity: 98% CAS No.: 56278-50-3 Molecular Formula: C9H6N2S Formula Weight: 174.23 Melting Point: 99-103° Beilstein Reference: 128460 UN No.: 3439 MDL No.: MFCD00051633

Formula: C₉H₆N₂S
MW: 174.23 g/mol
Melting Pt: 99…103 °C
Storage Temperature: Ambient
MDL Number: MFCD00051633
CAS Number: 56278-50-3
UN: 3439
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR