3,4-(Methylenedioxy)phenylacetic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (1,3-Benzodioxol-5-yl)acetic acid

A11229-14 A11229-22 A11229-06
AAA11229-14EA 286 USD
AAA11229-14 AAA11229-22 AAA11229-06
3,4-(Methylenedioxy)phenylacetic acid 98%
3,4-(Methylenedioxy)phenylacetic acid

Purity: 98% CAS No.: 2861-28-1 Molecular Formula: C9H8O4 Formula Weight: 180.16 Melting Point: 127-130° Beilstein Reference: 177739

Formula: C₉H₈O₄
MW: 180.16
Melting Pt: 127…130 °C
Storage Temperature: Ambient
MDL Number: MFCD00014576
CAS Number: 2861-28-1
EINECS: 220-679-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR