Hemin chloride (from porcine) 97+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Chlorohemin, 1,3,5,8-Tetramethyl-2,4-divinylporphine-6,7-dipropionic acid ferrichloride, Chloroprotoporphyrin IX iron(III), Haemin, Hemin, Ferriprotoporphyrin IX chloride

A11165-06 A11165-14 AAA11165-03
AAA11165-06EA 348.6 USD
AAA11165-06 AAA11165-14 AAA11165-03
Hemin chloride (from porcine) 97+%
Hemin chloride

Stimulates synthesis of globulin


Hemin is used in cellular protection and control mechanism. It stimulates the synthesis of globulin. It is also used to study the bacterial strains and growth conditions, in the short-term colony assays and cell transfections. Further, it is used to treat the symptoms of various porphyrias. In addition, it is used in the preparation of agar medium for the bacteria culture.

Formula: C₃₄H₃₂ClFeN₄O₄
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00010726
CAS Number: 16009-13-5
EINECS: 240-140-1
Merck Index: 14,4644

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR