4-Methylbenzoylacetonitrile 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10950-03
A10950-06
A10950-14
AAA10950-03EA
26.6
USD
AAA10950-03
AAA10950-06
AAA10950-14
4-Methylbenzoylacetonitrile 97%
4-Methylbenzoylacetonitrile
Purity: 97%
CAS No.: 7391-28-8
Molecular Formula: C10H9NO
Formula Weight: 159.19
Melting Point: 99-104°
Beilstein Reference: 742923
MDL No.: MFCD00067895
Formula:
C₁₀H₉NO MW: 159.188 g/mol Melting Pt: 99…104 °C Storage Temperature: Ambient |
MDL Number:
MFCD00067895 CAS Number: 7391-28-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...