3-Chloro-4-fluorophenylhydrazine hydrochloride 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(3-Chloro-4-fluorophenyl)hydrazine hydrochloride
A10175-14
A10175-06
AAA10175-22EA
1297.19
USD
AAA10175-22
AAA10175-14
AAA10175-06
3-Chloro-4-fluorophenylhydrazine hydrochloride 98%
3-Chloro-4-fluorophenylhydrazine hydrochloride
Purity: 98%
CAS No.: 175135-74-7
Molecular Formula: C6H62FN2.HCl
Formula Weight: 197.04
Melting Point: ca 210° dec.
MDL No.: MFCD00052267
Formula:
C₆H₇Cl₂FN₂ MW: 197.04 g/mol Melting Pt: ca 210 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00052267 CAS Number: 175135-74-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...