Glove Bag, Polyethylene, Thermo Scientific
Supplier: Thermo Scientific Chemicals
091717-LL 092717-LL 092727-LL 093727-LK 093727-LL 093737-LL 091717-LK 092717-LK 092727-LK 093737-LK
AA91717-LLEA 399.91 USD
AA91717-LL AA92717-LL AA92727-LL AA93727-LK AA93727-LL AA93737-LL AAA91717-LK AAA92717-LK AAA92727-LK AAA93737-LK
Glove Bag, Polyethylene, Thermo Scientific
Glove Boxes and Glove Bags
Polyethylene glove bag is used for laboratory use and field sampling
Suitable for handling the air and moisture sensitive, high purity chemicals and materials.
Información de suministro: Bag includes closure, tubing coupler and cotton gloves.
Order Now
SPECIFICATIONS
Environmentally Preferable