Nickel molybdenum oxide ≥98%

Supplier: Thermo Scientific Chemicals
Danger

089938-14 089938-22
AA89938-14EA 74.08 USD
AA89938-14 AA89938-22
Nickel molybdenum oxide ≥98%
Nickel molybdenum oxide

Purity: 98% CAS No.: 14177-55-0 Molecular Formula: NiMoO4 Formula Weight: 218.65 Physical Form: Powder Odor: Odorless MDL No.: MFCD00016255

Formula: NiMoO₄
MW: 218.65 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00016255
CAS Number: 14177-55-0
EINECS: 238-034-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR