Zirconium(IV) isopropoxide isopropanol complex
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Zirconium(IV) i-propoxide (isopropanol adduct)
086118-09
086118-18
AA86118-09EA
107
USD
AA86118-09
AA86118-18
Zirconium(IV) isopropoxide isopropanol complex
Zirconium(IV) isopropoxide isopropanol complex
Soluble in hot Isopropanol, warm hexane and toluene. Tends to precipitate over time
CAS No.: 14717-56-7
Molecular Formula: Zr[OCH(CH3)2]4.(CH3)2CHOH
Formula Weight: 387.67
Physical Form: Crystalline
Sensitivity: Moisture Sensitive
Boiling Point: dec.
Solubility: Soluble in hot Isopropanol, warm hexane and toluene. Tends to precipitate over time
UN No.: 3181
MDL No.: MFCD00075157
Formula:
C₁₅H₃₆O₅Zr MW: 387.672 g/mol Boiling Pt: Dec. Storage Temperature: Ambient |
MDL Number:
MFCD00075157 CAS Number: 14717-56-7 EINECS: 218-522-4 UN: 3181 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...