Tetraammineplatinum(II) hydroxide hydrate (≥58% Pt)
Supplier: Thermo Scientific Chemicals
|
Warning
|
046319-03
046319-06
AA46319-03EA
173
USD
AA46319-03
AA46319-06
Tetraammineplatinum(II) hydroxide hydrate (≥58% Pt)
Tetraammineplatinum(II) hydroxide hydrate
CAS No.: 312695-70-8
Molecular Formula: H14N4O2Pt.xH2O
Formula Weight: 297.23(anhy)
Physical Form: Powder
Appearance: Off-white
MDL No.: MFCD00151253
Formula:
H₁₄N₄O₂Pt·xH₂O Storage Temperature: Ambient |
MDL Number:
MFCD00151253 CAS Number: 312695-70-8 EINECS: 253-823-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...