Copper(II) ethoxide ≥98%

Supplier: Thermo Scientific Chemicals
Warning

046316-06 046316-14
AA46316-03EA 44.08 USD
AA46316-03 AA46316-06 AA46316-14 AA46316-22
Copper(II) ethoxide ≥98%
Copper(II) ethoxide

Purity: 98% CAS No.: 2850-65-9 Molecular Formula: C4H10CuO2 Formula Weight: 153.67 Physical Form: Powder, Packaged under Argon Sensitivity: Air and Moisture Sensitive Appearance: Green Melting Point: ca 120° dec. UN No.: 3181 MDL No.: MFCD00015650

Formula: C₄H₁₀CuO₂
MW: 153.67 g/mol
Melting Pt: ∼ 120 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00015650
CAS Number: 2850-65-9
UN: 3181
ADR: 4.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR