Tetrabutylammonium hydroxide 1.0 M in aqueous solution

Supplier: Thermo Scientific Chemicals
Danger

046315-AE 046315-AP
AA46315-AEEA 143 USD
AA46315-AE AA46315-AP
Tetrabutylammonium hydroxide 1.0 M in aqueous solution
Tetrabutylammonium hydroxide

CAS No.: 2052-49-5 Molecular Formula: C16H37NO Formula Weight: 259.48 Flash Point: None Physical Form: Liquid Sensitivity: Air Sensitive UN No.: 3267 MDL No.: MFCD00009425

Formula: C₁₆H₃₇NO
MW: 259.48 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00009425
CAS Number: 2052-49-5
EINECS: 218-147-6
UN: 3267
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR