Platinum 5% on activated carbon wetted with ca. 50% water, powder, reduced Type B105022-5

Supplier: Thermo Scientific Chemicals

046306-06 046306-14 046306-22
AA46306-06EA 113 USD
AA46306-06 AA46306-14 AA46306-22
Platinum 5% on activated carbon wetted with ca. 50% water, powder, reduced Type B105022-5
Platinum

CAS No.: 7440-06-4 Physical Form: 900-1100 m²/g Appearance: Dark grey to black Odor: Odorless MDL No.: MFCD00011179


Información para pedidos: Sold on a dry weight basis. Unit weight excludes water weight.


Precaución: Incompatible with acids and oxidizing agents

Formula: Pt
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS Number: 7440-06-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR