Aluminium oxide superactivated acidic Grade I

Supplier: Thermo Scientific Chemicals

046065-30 046065-A1
AA46065-30EA 75 USD
AA46065-30 AA46065-A1
Aluminium oxide superactivated acidic Grade I
Aluminium oxide superactivated acidic

CAS No.: 1344-28-1 Molecular Formula: Al2O3 Formula Weight: 101.96 Physical Form: 50-200 Micron APS Powder Refractive Index: 1.765 Appearance: White Odor: Odorless Melting Point: 2045° Boiling Point: 2980° MDL No.: MFCD00003424

Formula: Al₂O₃
MW: 101.96 g/mol
Boiling Pt: 2980 °C
Melting Pt: 2045 °C
Storage Temperature: Ambient
MDL Number: MFCD00003424
CAS Number: 1344-28-1
EINECS: 215-691-6
Merck Index: 14,356

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR