Bis[1,2,3,4,5-pentaphenyl-1'-(di-tert-butylphosphino)ferrocene]palladium(0)
Supplier: Thermo Scientific Chemicals
|
Warning
|
045913-03
045913-MD
AA45913-03EA
801
USD
AA45913-03
AA45913-MD
Bis[1,2,3,4,5-pentaphenyl-1'-(di-tert-butylphosphino)ferrocene]palladium(0)
Bis[1,2,3,4,5-pentaphenyl-1'-(di-tert-butylphosphino)ferrocene]palladium(0)
CAS No.: 565441-56-7
Molecular Formula: C96H94FeP2Pd
Formula Weight: 1527.86
Physical Form: Crystalline
Sensitivity: Air Sensitive
Appearance: Off-white to pink
MDL No.: MFCD15071401
Formula:
C₉₆H₉₄FeP₂Pd MW: 1527.86 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD15071401 CAS Number: 565441-56-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...