Poly(3-hexylthiophene-2,5-diyl) regiorandom
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
P3HT, Poly(3-hexylthiophene), Poly[3-hexylthiophene-2,5-diyl]
045906-MC
045906-ME
AA45906-04EA
1506.65
USD
AA45906-04
AA45906-MC
AA45906-ME
Poly(3-hexylthiophene-2,5-diyl) regiorandom
Poly(3-hexylthiophene-2,5-diyl)
CAS No.: 104934-50-1
Physical Form: Powder/chunks
Sensitivity: Light Sensitive
Appearance: Brown
MDL No.: MFCD00217686
MW:
196.352 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00217686 CAS Number: 104934-50-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...