Zeolite ZSM-5, ammonium
Supplier: Thermo Scientific Chemicals
045883-14
045883-22
045883-36
AA45883-14EA
33.7
USD
AA45883-14
AA45883-22
AA45883-36
Zeolite ZSM-5, ammonium
Zeolite ZSM-5, ammonium
Zeolite ZSM-5 is used in hydrocarbon isomerization, the alkylation of hydrocarbons, and as a catalyst support
CAS No.: 1318-02-1
Molecular Formula: SiO2:Al2O3
Physical Form: Powder, S.A. 400 m2/g, 200-400:1 mole ratio
Appearance: White
MDL No.: MFCD00132601
Applications: Zeolite ZSM-5 is used in hydrocarbon isomerization, the alkylation of hydrocarbons, and as a catalyst support
Formula:
SiO₂:Al₂O₃ Storage Temperature: Ambient |
MDL Number:
MFCD00132601 CAS Number: 1318-02-1 EINECS: 215-283-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...