Zeolite ZSM-5, ammonium
Supplier: Thermo Scientific Chemicals
045882-14
045882-22
045882-36
AA45882-14EA
33.7
USD
AA45882-14
AA45882-22
AA45882-36
Zeolite ZSM-5, ammonium
Zeolite ZSM-5, ammonium
Zeolite ZSM-5 is used in hydrocarbon isomerization, the alkylation of hydrocarbons, and as a catalyst support
CAS No.: 1318-02-1
Molecular Formula: SiO2:Al2O3
Physical Form: Powder, S.A. 425 m2/g, 80:1 mole ratio
Appearance: White
MDL No.: MFCD00132601
Applications: Zeolite ZSM-5 is used in hydrocarbon isomerization, the alkylation of hydrocarbons, and as a catalyst support
Formula:
SiO₂:Al₂O₃ Storage Temperature: Ambient |
MDL Number:
MFCD00132601 CAS Number: 1318-02-1 EINECS: 215-283-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...