Zeolite ZSM-5, ammonium

Supplier: Thermo Scientific Chemicals

045879-14 045879-22 045879-36
AA45879-14EA 31.7 USD
AA45879-14 AA45879-22 AA45879-36
Zeolite ZSM-5, ammonium
Zeolite ZSM-5, ammonium

Zeolite ZSM-5 is used in hydrocarbon isomerization, the alkylation of hydrocarbons, and as a catalyst support


CAS No.: 1318-02-1 Molecular Formula: SiO2:Al2O3 Physical Form: Powder, S.A. 425 m2/g, 23:1 mole ratio Appearance: White MDL No.: MFCD00132601


Applications: Zeolite ZSM-5 is used in hydrocarbon isomerization, the alkylation of hydrocarbons, and as a catalyst support

Formula: SiO₂:Al₂O₃
Storage Temperature: Ambient
MDL Number: MFCD00132601
CAS Number: 1318-02-1
EINECS: 215-283-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR