Zeolite mordenite, sodium
Supplier: Thermo Scientific Chemicals
045876-14
045876-22
045876-36
AA45876-14EA
31.3
USD
AA45876-14
AA45876-22
AA45876-36
Zeolite mordenite, sodium
Zeolite mordenite, sodium
Zeolite mordenite is used for the acid-catalyzed isomerization of alkanes and aromatics
CAS No.: 1318-02-1
Molecular Formula: SiO2:Al2O3
Physical Form: Powder, S.A. 425 m2/g, 13:1 mole ratio
Appearance: White
MDL No.: MFCD00132601
Applications: Zeolite mordenite is used for the acid-catalyzed isomerization of alkanes and aromatics
Formula:
SiO₂:Al₂O₃ Storage Temperature: Ambient |
MDL Number:
MFCD00132601 CAS Number: 1318-02-1 EINECS: 215-283-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...