Zeolite β, ammonium
Supplier: Thermo Scientific Chemicals
045873-14
045873-22
045873-36
AA45873-14EA
31.7
USD
AA45873-14
AA45873-22
AA45873-36
Zeolite β, ammonium
Zeolite beta, ammonium
Zeolite beta is used in isomerization of waxes, and Friedel Crafts reactions (alkylation and acylation)
CAS No.: 1318-02-1
Molecular Formula: SiO2:Al2O3
Physical Form: Powder, S.A. 680 m2/g, 25:1 mole ratio
Appearance: White
MDL No.: MFCD00132601
Applications: Zeolite beta is used in isomerization of waxes, and Friedel Crafts reactions (alkylation and acylation)
Formula:
SiO₂:Al₂O₃ Storage Temperature: Ambient |
MDL Number:
MFCD00132601 CAS Number: 1318-02-1 EINECS: 215-283-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...