Zeolite Y, hydrogen
Supplier: Thermo Scientific Chemicals
Zeolite Y is used as a cracking catalyst in petroleum refinery catalytic cracking units to convert high-boiling fractions of petroleum crude to more valuable gasoline, diesel and other products
CAS No.: 1318-02-1
Molecular Formula: SiO2:Al2O3
Physical Form: Powder, S.A. 780 m2/g, 80:1 mole ratio
Appearance: White
MDL No.: MFCD00132601
Applications: Zeolite Y is used as a cracking catalyst in petroleum refinery catalytic cracking units to convert high-boiling fractions of petroleum crude to more valuable gasoline, diesel and other products
Formula:
SiO₂:Al₂O₃ Storage Temperature: Ambient |
MDL Number:
MFCD00132601 CAS Number: 1318-02-1 EINECS: 215-283-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...