Bis(di-tert-butylphenylphosphine)palladium(0) 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
045853-03
045853-MD
AA45853-03EA
618
USD
AA45853-03
AA45853-MD
Bis(di-tert-butylphenylphosphine)palladium(0) 98%
Bis(di-tert-butylphenylphosphine)palladium(0)
CAS No.: 52359-17-8
Molecular Formula: C28H46P2Pd
Formula Weight: 551.04
Physical Form: Crystalline
Sensitivity: Air Sensitive
Appearance: Off-white
MDL No.: MFCD15071400
Formula:
C₂₈H₄₆P₂Pd Storage Temperature: Ambient |
MDL Number:
MFCD15071400 CAS Number: 52359-17-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...